


(S)-(+)-2-Octyl isocyanate 95%
Catalog # L20245.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:155.24 g/mol
Boiling Pt:68 °C (5 mmHg)
Density:0,826
ADR:6.1,III
:GHS06
:GHS08
:GHS07
CAS Number:745783-87-3
Formula:C₉H₁₇NO
MDL Number:MFCD05664089