DL-Malic acid disodium salt for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:676-46-0
:
Formula:C₄H₄Na₂O₅
MDL Number:MFCD00012466
:M-Clarity™ quality level = MQ200
EINECS:211-627-6
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:178.05 g/mol
Specification Test Results
Frequently Bought Together