2',7'-Dichlorofluorescein
Catalog # MOLEM27741374
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:401.20 g/mol
EINECS:200-968-6
CAS Number:76-54-0
Formula:C₂₀H₁₀Cl₂O₅
MDL Number:MFCD00005047