
8-Octanelactam ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:935-30-8
:
Formula:C₈H₁₅NO
MDL Number:MFCD00003273
:141.21 g/mol
Melting Pt:72…75 °C
EINECS:213-300-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together