4-(2-Pyridylazo)resorcinol monosodim salt monohydrate, Supelco®
Catalog # 1.07533.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Solubility:38 g/l (20 °C)
- Molar Mass:255.21 g/mol
- Bulk Density:630 kg/m3
- pH:9.8 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100
:255.21 g/mol (237.19 anhy)
EINECS:236-339-8
CAS Number:16593-81-0
Formula:C₁₁H₈N₃NaO₂·1H₂O
MDL Number:MFCD00150712