


Sodium-4-hydroxymercuriobenzoate for synthesis, Sigma-Aldrich®
Catalog # 8.20308.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:360.7 g/mol
Specification Test Results
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:205-340-5
UN:2025
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:138-85-2
Formula:C₇H₅HgNaO₃