Methyltributylammonium chloride ≥75% in aqueous solution Aliquat 175
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56375-79-2
:
Formula:C₁₃H₃₀ClN
MDL Number:MFCD00011847
:235.84 g/mol
Boiling Pt:84…85 °C (101 mmHg)
Melting Pt:–33 °C
Density:0,964
Storage Temperature:Ambient
EINECS:260-135-8
:GHS07
Related Information
Frequently Bought Together