



Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w)
:22722-98-1
:
:C₆H₁₆AlNaO₄
:MFCD00011631
:202.16 g/mol
:1,036 g/cm³
:4 °C
:Ambient
:245-178-2
:3399
:4.3,I
:GHS07
:GHS08
:GHS05
:GHS02
- Appearance:Clear to slightly hazy colorless to amber Viscous liquid
- Assay:68.0 to 72.0 %
- Comments:Liquid
- D-Enrichment:≥99.4 %
Frequently Bought Together