Order Entry
Northern Ireland
ContactUsLinkComponent
 
N,O-Dimethylhydroxylamine hydrochloride
Catalog # MOLEM17470360
Supplier:  Molekula
CAS Number:  
N,O-Dimethylhydroxylamine hydrochloride
Catalog # MOLEM17470360
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :97.55 g/mol
EINECS:229-642-1
CAS Number:6638-79-5
Formula:C₂H₈ClNO
MDL Number:MFCD00012485