
N,O-Dimethylhydroxylamine hydrochloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6638-79-5
:
Formula:C₂H₈ClNO
MDL Number:MFCD00012485
:97.55 g/mol
:M-Clarity™ quality level = MQ200
EINECS:229-642-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:112 - 116 °C
- Molar Mass:97.54 g/mol
Specification Test Results
Frequently Bought Together