


(S)-(-)-1-(4-Methoxyphenyl)ethyl isocyanate 95%
Catalog # L20240.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:177.20 g/mol
ADR:6.1,III
:GHS06
:GHS08
:GHS07
CAS Number:745783-83-9
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD05664085