
2-Bromo-1,3-diethylbenzene ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65232-57-7
:
Formula:C₁₀H₁₃Br
MDL Number:MFCD00017770
:213.12 g/mol
Boiling Pt:78…80 °C (2 mmHg)
Density:1,264
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together