Carminic acid, Supelco®
Catalog # 11298-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:215-023-3
CAS Number:1260-17-9
Formula:C₂₂H₂₀O₁₃
MDL Number:MFCD00167028