Methyl phenylglyoxylate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15206-55-0
:
Formula:C₉H₈O₃
MDL Number:MFCD00008443
:164.1600 g/mol
Boiling Pt:246…248 °C (760 mmHg)
Melting Pt:16 °C
Density:1,15 g/cm³
Flash Pt:77 °C
Storage Temperature:Ambient
EINECS:239-263-3
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5250 to 1.5280 (20°C, 589 nm)
- GC:≥98.5 %
- Substance darkened by H2SO4:Passes test
Specification Test Results
Frequently Bought Together