N-Isopropylpropylamine 96%
Catalog # H50256.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:21968-17-2
:
Formula:C₆H₁₅N
:101.19 g/mol
Storage Temperature:Ambient
EINECS:244-688-2
ADR:3,II
:GHS02
:GHS05
MDL Number:MFCD00027095