Phthalhydrazide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1445-69-8
:
Formula:C₈H₆N₂O₂
MDL Number:MFCD00006888
:162.15 g/mol
Melting Pt:342…345 °C
Storage Temperature:Ambient
EINECS:215-893-4
Related Information
Frequently Bought Together