Pyridoxal-5'-phosphate monohydrate
Catalog # MOLE26067672-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:265.17 g/mol (247.15 anhy)
Storage Temperature:Freezer
CAS Number:41468-25-1
Formula:C₈H₁₀NO₆P·1H₂O
MDL Number:MFCD00149414