Order Entry
Northern Ireland
ContactUsLinkComponent
L(-)-Carnitine ≥98.0% for synthesis pH 6.5 - 8.5 (50 g/l, H₂O), Sigma-Aldrich®
L(-)-Carnitine ≥98.0% for synthesis pH 6.5 - 8.5 (50 g/l, H₂O), Sigma-Aldrich®
Catalog # 8.40092.0025
Supplier:  Merck
CAS Number:  
L(-)-Carnitine ≥98.0% for synthesis pH 6.5 - 8.5 (50 g/l, H₂O), Sigma-Aldrich®
Catalog # 8.40092.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    800 g/l (20 °C)
  • Melting Point:
    190 - 195 °C decomposes
  • Molar Mass:
    161.2 g/mol
  • Bulk Density:
    636 kg/m3
  • Density:
    0.64 g/cm3
  • pH:
    6.5 - 8.5 (50 g/l, H2O)
  • Ignition Point:
    365 °C

Specification Test Results

About this item

 :
 :161.20 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:208-768-0
CAS Number:541-15-1
Formula:C₇H₁₅NO₃
MDL Number:MFCD00038747