DL-Carnitine hydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.41774.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:196 - 197 °C decomposes
- Molar Mass:197.66 g/mol
- pH:1.8 - 2.5 (100 g/l, H2O)
Specification Test Results
About this item
Synonyms: 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride
:
:197.66 g/mol
EINECS:207-309-1
CAS Number:461-05-2
Formula:C₇H₁₆ClNO₃
MDL Number:MFCD00011904
:M-Clarity™ quality level = MQ200