
Potassium tetrakis(4-chlorophenyl)borate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14680-77-4
:
Formula:C₂₄H₁₆BCl₄K
MDL Number:MFCD00043105
:496.12 g/mol
Melting Pt:>300 °C
EINECS:238-723-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together