Order Entry
Northern Ireland
ContactUsLinkComponent
Bis(4-fluorophenyl)methane for synthesis
Bis(4-fluorophenyl)methane for synthesis
Catalog # 8.40056.0010
Supplier:  Merck
CAS Number:  
Bis(4-fluorophenyl)methane for synthesis
Catalog # 8.40056.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Molar Mass:
    204.22 g/mol
  • Boiling Point:
    265 °C
  • Density:
    1.16 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :204.22 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:207-274-2
CAS Number:457-68-1
Formula:C₁₃H₁₀F₂
MDL Number:MFCD00000360