
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14470-28-1
:
Formula:C₂₀H₁₇ClO
MDL Number:MFCD00000814
:308.8 g/mol
Melting Pt:121…125 °C
EINECS:238-463-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together