Alcian blue 8GX
Catalog # MOLE59508864-10G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:1298.86 g/mol
EINECS:251-705-7
:GHS07
Storage Temperature:Ambient
CAS Number:33864-99-2
:75881-23-1
Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:MFCD00010720