Cumene hydroperoxide 80% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80-15-9
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00002129
:152.20 g/mol
Melting Pt:–30 °C
Density:1,06 g/cm³
Flash Pt:83 °C
Storage Temperature:Refrigerator
EINECS:201-254-7
UN:3109
ADR:5.2,
:GHS02
:GHS06
:GHS08
:GHS05
:GHS09
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Titration Iodometric:80 to 85 %
- Color scale:≤450 APHA
Specification Test Results
Frequently Bought Together