
3,5-Dibromo-2,6-dimethoxypyridine ≥99%
Catalog # H27598.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:16727-44-9
:
Formula:C₇H₇Br₂NO₂
:296.95 g/mol
Melting Pt:85…87 °C
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00955954