Order Entry
Northern Ireland
ContactUsLinkComponent
 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -18 °C
  • Molar Mass:
    153.26 g/mol
  • Boiling Point:
    114 °C (13 hPa)
  • Refractive Index:
    1.4313 (15 °C, 589 nm)
  • Density:
    0.82 g/cm3 (20 °C)
  • Synonyms:
    Capric acid nitrile
  • Hill Formula:
    C10H19N
  • Chemical Formula:
    CH3(CH2)8CN

Specification Test Results

About this item

Synonyms: Decanonitrile
 :
 :153.27 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:217-830-6
CAS Number:1975-78-6
Formula:C₁₀H₁₉N
MDL Number:MFCD00039505