Organic reference standard, Dibenz[a,h]anthracene 100 µg/ml in acetonitrile
Catalog # PROONE5039
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,5 mL
- CAS No.:53-70-3
- Description:Dibenz[a,h]anthracene 100 µg/ml in acetonitrile
- Formula:C₂₂H₁₄
- Description:Dibenz[a,h]anthracene