Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminoethyl dihydrogen phosphate
2-Aminoethyl dihydrogen phosphate
2-Aminoethyl dihydrogen phosphate
2-Aminoethyl dihydrogen phosphate
MOLE89847060-25G
:
Molekula
:
2-Aminoethyl dihydrogen phosphate
MOLE89847060-25G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
141.0630 g/mol
:
213-988-5
:
3261
:
8,II
:
1071-23-4
:
C₂H₈NO₄P
:
MFCD00008178