
2-Aminoethyl dihydrogen phosphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1071-23-4
:
Formula:C₂H₈NO₄P
MDL Number:MFCD00008178
:141.0630 g/mol
Melting Pt:241…243 °C
Storage Temperature:Freezer
EINECS:213-988-5
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together