
(4-Chlorobenzyl)triphenylphosphonium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1530-39-8
:
Formula:C₂₅H₂₁Cl₂P
MDL Number:MFCD00041533
:423.32 g/mol
Melting Pt:286…291 °C
EINECS:216-228-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together