(4-Bromobenzyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51044-13-4
:
Formula:C₂₅H₂₁Br₂P
MDL Number:MFCD00051922
:512.24 g/mol
Melting Pt:270…273 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together