Lutetium(III) acetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207500-05-8
:18779-08-3
Formula:C₆H₉LuO₆·xH₂O
MDL Number:MFCD00150119
:352.11 g/mol (anhy)
Storage Temperature:Ambient
EINECS:242-566-3
Related Information
Frequently Bought Together