Order Entry
Northern Ireland
ContactUsLinkComponent
 
N-Acetyl-DL-valine for synthesis, Sigma-Aldrich®
  8.14599.0050
 :  Merck
 :  
N-Acetyl-DL-valine for synthesis, Sigma-Aldrich®
  8.14599.0050
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

 

 

  • Solubility:
    37 g/l (25 °C)
  • Melting Point:
    146 - 149 °C
  • Molar Mass:
    159.18 g/mol
  • Hill Formula:
    C7H13NO3

 

 

 :
 :M-Clarity™ quality level = MQ200
 :159.2 g/mol
 :221-321-4
 :3067-19-4
 :C₇H₁₃NO₃
 :MFCD00066065