Order Entry
Northern Ireland
ContactUsLinkComponent
 
Isobutyl stearate for synthesis, Sigma-Aldrich®
 :  Merck
 
Isobutyl stearate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Melting Point:
    28.9 °C
  • Density:
    0.85 g/cm3 (20 °C)

 

 

Frequently Bought Together