Bovine trypsin (from pancreas)
786-262
:
- Pack type:Glass bottle for solids
- Pk:1 g
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Bovine
- Environmentally Preferable:
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
- Purity:1X crystallized