4-Amino-1H-indazole
Catalog # APOSOR16942-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:133.15 g/mol
CAS Number:41748-71-4
Formula:C₇H₇N₃
MDL Number:MFCD03305710