Order Entry
Northern Ireland
ContactUsLinkComponent
(Trifluoromethyl)trimethylsilane 0.5M in THF, AcroSeal®
(Trifluoromethyl)trimethylsilane 0.5M in THF, AcroSeal®
Catalog # 429171000.
CAS Number:  
undefined
(Trifluoromethyl)trimethylsilane 0.5M in THF, AcroSeal®
Catalog # 429171000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    0.45 to 0.55 M

Specification Test Results

About this item

 :
 :142.2 g/mol
Boiling Pt:40 °C (760 mmHg)
Density:0,895 g/cm³
Flash Pt:–20 °C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
 :GHS07
 :GHS02
 :GHS08
CAS Number:81290-20-2
Formula:C₄H₉F₃Si
MDL Number:MFCD00145454