Alizarin red S (sodium salt) 1% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:130-22-3
:93982-72-0
Formula:C₁₄H₇NaO₇S
MDL Number:MFCD00013049
:342.26 g/mol
Storage Temperature:Ambient
EINECS:204-981-8
Related Information
Frequently Bought Together