Soy bean trypsin inhibitor
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- CAS No.:9035-81-8
- Species:Soy bean
- Storage conditions:Freezer
- Protein/peptide name:Trypsin inhibitor
Specifications
Frequently Bought Together