Leupeptin hemisulphate
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- CAS No.:103476-89-7
- Storage conditions:Freezer
- MDL No.:MFCD00037012
- Protein synonyms:(Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
- Protein/peptide name:Leupeptin hemisulphate
- Molecular formula:C₄₀H₇₈N₁₂O₁₂S
Specifications
Frequently Bought Together