Order Entry
Northern Ireland
ContactUsLinkComponent
Antipain Dihydrochloride
Antipain Dihydrochloride
Certificates
Antipain Dihydrochloride

Specifications

  • CAS No.:
    37682-72-7
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00135957
  • Protein/peptide name:
    Antipain dihydrochloride
  • Molecular formula:
    C₂₇H₄₄N₁₀O₆ · 2HCl

Specifications

Frequently Bought Together