Order Entry
Northern Ireland
ContactUsLinkComponent
2'-Deoxycytidine hydrochloride
2'-Deoxycytidine hydrochloride
Catalog # MOLE14554862-1G
Supplier:  Molekula
CAS Number:  
2'-Deoxycytidine hydrochloride
Catalog # MOLE14554862-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
 :
 :263.68 g/mol
Storage Temperature:Ambient
EINECS:223-639-9
CAS Number:3992-42-5
Formula:C₉H₁₄ClN₃O₄
MDL Number:MFCD00012840