2'-Deoxyguanosine monohydrate
Catalog # MOLE15163303-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:285.26 g/mol
Storage Temperature:Ambient
EINECS:213-505-8
CAS Number:312693-72-4
Formula:C₁₀H₁₃N₅O₄·H₂O
MDL Number:MFCD00150760