
Tungsten(V) ethoxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26143-11-3
:
Formula:C₁₀H₂₅O₅W
MDL Number:MFCD00078041
:409.15 g/mol
Boiling Pt:110…115 °C (1 mmHg)
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together