

Tin(II) ethoxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14791-99-2
:
Formula:C₄H₁₀O₂Sn
MDL Number:MFCD00015653
:208.83 g/mol
Melting Pt:∼ 200 °C (dec.)
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together