3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl)
Catalog # MOLE27393749
Supplier: Molekula
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:360.11 g/mol
EINECS:231-018-9
CAS Number:7411-49-6
Formula:C₁₂H₁₈Cl₄N₄
MDL Number:MFCD00012969