Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Catalog # MOLEM29628672
Supplier:  Molekula
CAS Number:  
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Catalog # MOLEM29628672
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :160.30 g/mol
EINECS:205-439-3
CAS Number:140-89-6
Formula:C₃H₅KOS₂
MDL Number:MFCD00004931