Creatine phosphate dipotassium salt ≥98% (by TLC), Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
Creatine phosphate dipotassium salt ≥98% (by TLC), Millipore®
Supplier: Merck Millipore (Calbiochem)
About this item
CAS Number:18838-38-5
:
Formula:C₄H₈K₂N₃O₅P
Storage Temperature:Freezer
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Jars with loose enfolding lids, DURAN®
Supplier: DWK Life Sciences
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection