o-Cresolphthalein complexone indicator for metal titration
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: o-Cresolphthalein Complexone, Phthalein purple, Cresol phthalexone, Xylenolphthalein-bis(iminodiacetic acid), Phthalein complexone, Phthalate complexing agent [Chelation indicator], Metal phthaleine, o-Cresolphthalexon
CAS Number:2411-89-4
:
Formula:C₃₂H₃₂N₂O₁₂
MDL Number:MFCD00005911
Molecular Weight:636.62 g/mol
Melting Pt:∼ 198 °C (dec.)
Storage Temperature:Ambient
EINECS:219-318-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together