
Azure B (azure I)
:531-55-5
:
:C₁₅H₁₆ClN₃S
:MFCD00011935
:305.83 g/mol
:14,6058
:∼ 205 °C (dec.)
:208-511-2
:GHS08
:Ambient
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals